Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IBTK
Entrez Gene ID
25998
Gene Name
inhibitor of Bruton tyrosine kinase
Gene Aliases
BTBD26, BTKI
Chromosome Location
Chr.6: 82169987 - 82248294 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300906.1 NP_001287835.1 28 - 29 4442 143
NM_015525.3 NP_056340.2 28 - 29 4487 143
XM_006715453.2 XP_006715516.1 29 - 30 5150 143
XM_006715454.2 XP_006715517.1 29 - 30 5105 143
GenBank mRNA AB037838.2 - 27 - 28 4211 143
AF235049.1 - 5 - 6 906 143
AJ420588.1 - 3 - 4 371 143
AJ420593.1 - 4 - 5 391 143
AK123460.1 - 1 - 2 1207 143
AK126777.1 - 12 - 13 2352 143
AL050018.1 - 8 - 9 1027 143
BC027490.1 - 6 - 7 671 143
BC038244.2 - 11 - 12 1325 143
BC042171.1 - 7 - 8 870 143
BC113696.1 - 27 - 28 4016 143
BC113698.1 - 27 - 28 4016 143
BC143736.1 - 26 - 27 3413 143
BC143748.1 - 25 - 26 3983 143
DQ005633.1 - 28 - 29 4487 143
DQ005635.1 - 4 - 5 1017 143

View Full Product Details