The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300906.1 | NP_001287835.1 | 13 - 14 | 2841 | 120 |
NM_015525.3 | NP_056340.2 | 13 - 14 | 2841 | 120 | |
XM_006715453.2 | XP_006715516.1 | 13 - 14 | 3381 | 120 | |
XM_006715454.2 | XP_006715517.1 | 13 - 14 | 3381 | 120 | |
GenBank mRNA | AB037838.2 | - | 12 - 13 | 2565 | 120 |
BC113696.1 | - | 12 - 13 | 2370 | 120 | |
BC113698.1 | - | 12 - 13 | 2370 | 120 | |
BC143736.1 | - | 11 - 12 | 1767 | 120 | |
BC143745.1 | - | 12 - 13 | 2371 | 120 | |
BC143748.1 | - | 11 - 12 | 1767 | 120 | |
DQ005633.1 | - | 13 - 14 | 2841 | 120 | |
DQ005634.1 | - | 13 - 14 | 2841 | 120 |