The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009932.2 | NP_001009932.1 | 2 - 3 | 453 | 81 |
XM_005277829.4 | XP_005277886.1 | 2 - 3 | 587 | 81 | |
XM_011531122.2 | XP_011529424.1 | 1 - 2 | 582 | 81 | |
XM_017029332.1 | XP_016884821.1 | 2 - 3 | 1659 | 81 | |
GenBank mRNA | CN346700.1 | - | 2 - 3 | 471 | 81 |
X90392.1 | - | 1 - 2 | 616 | 81 | |
X90393.1 | - | 2 - 3 | 384 | 81 |