Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIMCH1
Entrez Gene ID
22998
Gene Name
LIM and calponin homology domains 1
Gene Aliases
LIMCH1A, LMO7B
Chromosome Location
Chr.4: 41359607 - 41700044 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112717.2 NP_001106188.1 10 - 11 1159 125
NM_001112718.2 NP_001106189.1 10 - 11 1159 125
NM_001112719.2 NP_001106190.1 5 - 6 944 125
NM_001112720.2 NP_001106191.1 4 - 5 908 125
NM_001289122.1 NP_001276051.1 9 - 10 1123 125
NM_001289124.1 NP_001276053.1 9 - 10 1168 125
NM_014988.3 NP_055803.2 10 - 11 1159 125
XM_005248057.1 XP_005248114.1 17 - 18 2791 125
XM_005248058.1 XP_005248115.1 17 - 18 2791 125
XM_005248060.1 XP_005248117.1 17 - 18 2791 125
XM_005248061.1 XP_005248118.1 17 - 18 2791 125
XM_005248067.2 XP_005248124.1 15 - 16 3665 125
XM_005248072.1 XP_005248129.1 10 - 11 1159 125
XM_005248074.1 XP_005248131.1 9 - 10 1123 125
XM_005248075.1 XP_005248132.1 9 - 10 1123 125
XM_006713996.1 XP_006714059.1 17 - 18 2791 125
XM_011513642.1 XP_011511944.1 17 - 18 2791 125
XM_011513643.1 XP_011511945.1 17 - 18 2791 125
XM_011513644.1 XP_011511946.1 17 - 18 2791 125
XM_011513645.1 XP_011511947.1 17 - 18 2791 125
XM_011513646.1 XP_011511948.1 17 - 18 2791 125
XM_011513648.1 XP_011511950.1 16 - 17 2548 125
XM_011513649.1 XP_011511951.1 12 - 13 2450 125
XM_011513651.2 XP_011511953.1 15 - 16 3670 125
XM_011513653.1 XP_011511955.1 10 - 11 1159 125
XM_011513654.1 XP_011511956.1 9 - 10 1123 125
XM_011513655.1 XP_011511957.1 4 - 5 782 125
XM_011513656.1 XP_011511958.1 5 - 6 818 125
XM_011513657.1 XP_011511959.1 4 - 5 782 125
XM_011513658.2 XP_011511960.1 9 - 10 1384 125
XM_017007894.1 XP_016863383.1 17 - 18 2791 125
XM_017007895.1 XP_016863384.1 17 - 18 2791 125
XM_017007896.1 XP_016863385.1 17 - 18 2791 125
XM_017007897.1 XP_016863386.1 17 - 18 2791 125
XM_017007898.1 XP_016863387.1 10 - 11 1159 125
XM_017007899.1 XP_016863388.1 10 - 11 1159 125
XM_017007900.1 XP_016863389.1 10 - 11 1159 125
XM_017007901.1 XP_016863390.1 10 - 11 1159 125
XM_017007902.1 XP_016863391.1 10 - 11 1159 125
XM_017007903.1 XP_016863392.1 5 - 6 818 125
XM_017007904.1 XP_016863393.1 5 - 6 818 125
XM_017007905.1 XP_016863394.1 5 - 6 818 125
GenBank mRNA AB029025.2 - 10 - 11 1157 125
AK294774.1 - 4 - 5 971 125
AK295784.1 - 3 - 4 497 125
AK298915.1 - 5 - 6 814 125
AK299297.1 - 4 - 5 778 125
AK299835.1 - 4 - 5 642 125
AK309846.1 - 4 - 5 850 125
AL831962.2 - 15 - 16 2546 125
BC023546.2 - 7 - 8 1012 125
BX537916.1 - 4 - 5 778 125
BX640692.1 - 3 - 4 538 125
CR749205.1 - 5 - 6 814 125
CR933645.1 - 10 - 11 1160 125
CR936601.1 - 9 - 10 1094 125
CR936610.1 - 9 - 10 1169 125
CR936658.1 - 10 - 11 1138 125
CR936661.1 - 9 - 10 1063 125
CR936664.1 - 10 - 11 1160 125

View Full Product Details