Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADPGK
Entrez Gene ID
83440
Gene Name
ADP dependent glucokinase
Gene Aliases
2610017G09Rik, ADP-GK
Chromosome Location
Chr.15: 72751367 - 72783785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031284.4 NP_112574.3 3 - 4 622 108
NR_023318.1 3 - 4 835 108
XM_006720712.3 XP_006720775.1 3 - 4 622 108
XM_006720713.3 XP_006720776.1 3 - 4 783 108
XM_011522104.2 XP_011520406.1 3 - 4 622 108
GenBank mRNA AF204270.1 - 2 - 3 211 108
AK075560.1 - 3 - 4 557 108
AK293282.1 - 3 - 4 314 108
AK294391.1 - 2 - 3 359 108
BC006112.1 - 3 - 4 564 108
BC044589.1 - 3 - 4 781 108

View Full Product Details