The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206747.1 | NP_001193676.1 | 1 - 2 | 330 | 54 |
NM_001233.4 | NP_001224.1 | 1 - 2 | 330 | 54 | |
GenBank mRNA | AB451285.1 | - | 1 - 2 | 149 | 54 |
AB451417.1 | - | 1 - 2 | 149 | 54 | |
AF035752.1 | - | 1 - 2 | 169 | 54 | |
AK311964.1 | - | 1 - 2 | 172 | 54 | |
BC005256.1 | - | 1 - 2 | 330 | 54 | |
BM762545.1 | - | 1 - 2 | 541 | 54 | |
BT007051.1 | - | 1 - 2 | 149 | 54 | |
CR407685.1 | - | 1 - 2 | 149 | 54 | |
DQ656068.1 | - | 11 - 12 | 1486 | 54 |