The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300918.1 | NP_001287847.1 | 3 - 4 | 1061 | 81 |
NM_003483.4 | NP_003474.1 | 3 - 4 | 1061 | 81 | |
GenBank mRNA | AB209853.1 | - | 3 - 4 | 623 | 81 |
AF533653.1 | - | - | 153 | 81 | |
U28749.1 | - | 3 - 4 | 1061 | 81 | |
X92518.1 | - | 3 - 4 | 1560 | 81 | |
Z31595.1 | - | 3 - 4 | 1062 | 81 |