The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138769.2 | NP_620124.1 | 3 - 4 | 296 | 81 |
XM_005255660.2 | XP_005255717.1 | 3 - 4 | 247 | 81 | |
XM_005255663.2 | XP_005255720.1 | 3 - 4 | 247 | 81 | |
XM_006720970.2 | XP_006721033.1 | 3 - 4 | 247 | 81 | |
XM_006720973.2 | XP_006721036.1 | 3 - 4 | 247 | 81 | |
XM_017023825.1 | XP_016879314.1 | 3 - 4 | 247 | 81 | |
XM_017023826.1 | XP_016879315.1 | 3 - 4 | 247 | 81 | |
XM_017023829.1 | XP_016879318.1 | 3 - 4 | 247 | 81 | |
XM_017023831.1 | XP_016879320.1 | 3 - 4 | 247 | 81 | |
XM_017023833.1 | XP_016879322.1 | 3 - 4 | 247 | 81 | |
XM_017023835.1 | XP_016879324.1 | 3 - 4 | 245 | 81 | |
XM_017023836.1 | XP_016879325.1 | 3 - 4 | 247 | 81 | |
XM_017023837.1 | XP_016879326.1 | 3 - 4 | 247 | 81 | |
XM_017023839.1 | XP_016879328.1 | 3 - 4 | 245 | 81 | |
XM_017023840.1 | XP_016879329.1 | 3 - 4 | 177 | 81 | |
XM_017023841.1 | XP_016879330.1 | 3 - 4 | 236 | 81 | |
XM_017023842.1 | XP_016879331.1 | 3 - 4 | 237 | 81 | |
GenBank mRNA | AJ517413.1 | - | 3 - 4 | 179 | 81 |
AY207375.1 | - | 3 - 4 | 293 | 81 | |
BC014942.1 | - | 3 - 4 | 246 | 81 | |
DB465261.1 | - | 3 - 4 | 297 | 81 |