The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015032.3 | NP_055847.1 | 16 - 17 | 1927 | 93 |
XM_005266298.4 | XP_005266355.1 | 16 - 17 | 1922 | 93 | |
XM_011534999.2 | XP_011533301.1 | 16 - 17 | 1922 | 93 | |
XM_011535000.2 | XP_011533302.1 | 16 - 17 | 1922 | 93 | |
XM_011535002.2 | XP_011533304.1 | 9 - 10 | 1024 | 93 | |
XM_017020448.1 | XP_016875937.1 | 16 - 17 | 2069 | 93 | |
XM_017020449.1 | XP_016875938.1 | 16 - 17 | 1837 | 93 | |
XM_017020450.1 | XP_016875939.1 | 16 - 17 | 1837 | 93 | |
XM_017020451.1 | XP_016875940.1 | 16 - 17 | 1837 | 93 | |
XM_017020452.1 | XP_016875941.1 | 16 - 17 | 1893 | 93 | |
XM_017020453.1 | XP_016875942.1 | 16 - 17 | 1922 | 93 | |
GenBank mRNA | AB023196.2 | - | 16 - 17 | 1849 | 93 |
AK299340.1 | - | 16 - 17 | 1803 | 93 | |
AL137201.1 | - | 16 - 17 | 1867 | 93 | |
U50533.1 | - | 6 - 7 | 678 | 93 | |
U95825.2 | - | 16 - 17 | 1806 | 93 |