The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015032.3 | NP_055847.1 | 27 - 28 | 3369 | 66 |
XM_005266298.4 | XP_005266355.1 | 27 - 28 | 3364 | 66 | |
XM_011534999.2 | XP_011533301.1 | 27 - 28 | 3364 | 66 | |
XM_011535000.2 | XP_011533302.1 | 27 - 28 | 3364 | 66 | |
XM_011535002.2 | XP_011533304.1 | 20 - 21 | 2466 | 66 | |
XM_017020448.1 | XP_016875937.1 | 27 - 28 | 3511 | 66 | |
XM_017020449.1 | XP_016875938.1 | 27 - 28 | 3279 | 66 | |
XM_017020450.1 | XP_016875939.1 | 27 - 28 | 3279 | 66 | |
XM_017020451.1 | XP_016875940.1 | 27 - 28 | 3279 | 66 | |
XM_017020452.1 | XP_016875941.1 | 27 - 28 | 3335 | 66 | |
XM_017020453.1 | XP_016875942.1 | 27 - 28 | 3364 | 66 | |
GenBank mRNA | AB023196.2 | - | 27 - 28 | 3291 | 66 |
AK128502.1 | - | 9 - 10 | 3151 | 66 | |
AK299340.1 | - | 27 - 28 | 3245 | 66 | |
AL137201.1 | - | 27 - 28 | 3309 | 66 | |
U95825.2 | - | 27 - 28 | 3248 | 66 |