The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002198.2 | NP_002189.1 | 7 - 8 | 897 | 74 |
XM_011543379.2 | XP_011541681.1 | 5 - 6 | 721 | 74 | |
GenBank mRNA | AB103081.1 | - | 6 - 7 | 663 | 74 |
AK314025.1 | - | 7 - 8 | 897 | 74 | |
BC009483.2 | - | 7 - 8 | 866 | 74 | |
BT019755.1 | - | 6 - 7 | 663 | 74 | |
BT019756.1 | - | 6 - 7 | 663 | 74 | |
CR541713.1 | - | 6 - 7 | 663 | 74 | |
KC209828.1 | - | 6 - 7 | 888 | 74 | |
KJ200632.1 | - | 7 - 8 | 779 | 74 | |
KJ200633.1 | - | 7 - 8 | 685 | 74 | |
X14454.1 | - | 7 - 8 | 860 | 74 |