The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002198.2 | NP_002189.1 | 8 - 9 | 955 | 76 |
XM_011543378.2 | XP_011541680.1 | 7 - 8 | 832 | 76 | |
XM_011543379.2 | XP_011541681.1 | 6 - 7 | 779 | 76 | |
GenBank mRNA | AK314025.1 | - | 8 - 9 | 955 | 76 |
BC009483.2 | - | 8 - 9 | 924 | 76 | |
BT019755.1 | - | 7 - 8 | 721 | 76 | |
BT019756.1 | - | 7 - 8 | 721 | 76 | |
CR541713.1 | - | 7 - 8 | 721 | 76 | |
KC209828.1 | - | 7 - 8 | 946 | 76 | |
KJ200632.1 | - | 8 - 9 | 837 | 76 | |
KJ200633.1 | - | 8 - 9 | 743 | 76 | |
KJ200634.1 | - | 7 - 8 | 714 | 76 | |
X14454.1 | - | 8 - 9 | 918 | 76 |