The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165257.1 | NP_001158729.1 | 9 - 10 | 1548 | 125 |
NM_013243.3 | NP_037375.2 | 10 - 11 | 1601 | 125 | |
GenBank mRNA | AF078851.1 | - | 10 - 11 | 1483 | 125 |
AF453583.1 | - | 10 - 11 | 1606 | 125 | |
AK075314.1 | - | 10 - 11 | 1460 | 125 | |
AK290175.1 | - | 10 - 11 | 1437 | 125 | |
AK296466.1 | - | 9 - 10 | 1387 | 125 | |
AY359093.1 | - | 10 - 11 | 1462 | 125 | |
BC009511.2 | - | 4 - 5 | 493 | 125 | |
BC014539.1 | - | 10 - 11 | 1237 | 125 |