The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286974.1 | NP_001273903.1 | 15 - 16 | 2031 | 113 |
NM_003798.3 | NP_003789.1 | 15 - 16 | 2031 | 113 | |
XM_005252291.3 | XP_005252348.1 | 15 - 16 | 1999 | 113 | |
XM_017015250.1 | XP_016870739.1 | 14 - 15 | 1751 | 113 | |
XM_017015251.1 | XP_016870740.1 | 14 - 15 | 1751 | 113 | |
GenBank mRNA | AB451283.1 | - | 16 - 17 | 1879 | 113 |
AB451415.1 | - | 16 - 17 | 1879 | 113 | |
AF006070.1 | - | 15 - 16 | 1921 | 113 | |
AF030233.1 | - | 15 - 16 | 1923 | 113 | |
AF080071.1 | - | 15 - 16 | 1918 | 113 | |
AF135021.1 | - | 15 - 16 | 1922 | 113 | |
AK022539.1 | - | 14 - 15 | 1713 | 113 | |
AK022834.1 | - | 15 - 16 | 1964 | 113 | |
AK022926.1 | - | 15 - 16 | 1936 | 113 | |
AK223230.1 | - | 15 - 16 | 1936 | 113 | |
AK314677.1 | - | 15 - 16 | 1935 | 113 | |
AL050016.1 | - | 8 - 9 | 743 | 113 | |
BC117208.1 | - | 15 - 16 | 1900 | 113 | |
BC117234.1 | - | 15 - 16 | 1900 | 113 | |
BU677451.1 | - | 4 - 5 | 113 | ||
U97067.1 | - | 15 - 16 | 1922 | 113 |