Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTNNAL1
Entrez Gene ID
8727
Gene Name
catenin alpha like 1
Gene Aliases
ACRP, CLLP, alpha-CATU
Chromosome Location
Chr.9: 108942569 - 109013594 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286974.1 NP_001273903.1 5 - 6 879 126
NM_003798.3 NP_003789.1 5 - 6 879 126
XM_005252291.3 XP_005252348.1 5 - 6 847 126
XM_017015250.1 XP_016870739.1 4 - 5 599 126
XM_017015251.1 XP_016870740.1 4 - 5 599 126
GenBank mRNA AF006070.1 - 5 - 6 769 126
AF030233.1 - 5 - 6 771 126
AF080071.1 - 5 - 6 766 126
AF135021.1 - 5 - 6 770 126
AK022539.1 - 4 - 5 561 126
AK022834.1 - 5 - 6 812 126
AK022926.1 - 5 - 6 784 126
AK223230.1 - 5 - 6 784 126
AK314677.1 - 5 - 6 783 126
BC117208.1 - 5 - 6 748 126
BC117234.1 - 5 - 6 748 126
U97067.1 - 5 - 6 770 126

View Full Product Details