Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKD1L2
Entrez Gene ID
114780
Gene Name
polycystin 1 like 2 (gene/pseudogene)
Gene Aliases
PC1L2
Chromosome Location
Chr.16: 81100879 - 81220370 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076780.1 NP_001070248.1 15 - 16 2587 136
NM_001278423.1 NP_001265352.1 4 - 5 681 136
NM_001278425.1 NP_001265354.1 4 - 5 681 136
NM_052892.3 NP_443124.3 15 - 16 2587 136
NR_126532.1 15 - 16 2587 136
GenBank mRNA AK098052.1 - 9 - 10 1169 136
AK127266.1 - 3 - 4 469 136
AY371495.1 - 4 - 5 643 136
BC004562.2 - 4 - 5 648 136
BC014157.2 - 4 - 5 648 136
CR627341.1 - 2 - 3 174 136

View Full Product Details