The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324444.1 | NP_001311373.1 | 3 - 4 | 334 | 62 |
NM_001324445.1 | NP_001311374.1 | 2 - 3 | 1107 | 62 | |
NM_001324446.1 | NP_001311375.1 | 2 - 3 | 1013 | 62 | |
NM_001324448.1 | NP_001311377.1 | 3 - 4 | 334 | 62 | |
NM_001324449.1 | NP_001311378.1 | 2 - 3 | 1107 | 62 | |
NM_001324450.1 | NP_001311379.1 | 3 - 4 | 334 | 62 | |
NM_001324451.1 | NP_001311380.1 | 2 - 3 | 1107 | 62 | |
NM_001324452.1 | NP_001311381.1 | 2 - 3 | 1107 | 62 | |
NM_001324453.1 | NP_001311382.1 | 2 - 3 | 1107 | 62 | |
NM_012091.4 | NP_036223.2 | 3 - 4 | 334 | 62 | |
GenBank mRNA | AK001687.1 | - | 3 - 4 | 267 | 62 |
AK296834.1 | - | 4 - 5 | 205 | 62 | |
AK314652.1 | - | 3 - 4 | 276 | 62 | |
BC002758.2 | - | 3 - 4 | 280 | 62 | |
DC395938.1 | - | 3 - 4 | 334 | 62 |