Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MKL2
Entrez Gene ID
57496
Gene Name
MKL1/myocardin like 2
Gene Aliases
MRTF-B, MRTFB, NPD001
Chromosome Location
Chr.16: 14071076 - 14266779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC047761.1 - 8 - 9 1080 69

View Full Product Details