The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164104.1 | NP_001157576.1 | 9 - 10 | 1542 | 152 |
NM_001164105.1 | NP_001157577.1 | 9 - 10 | 1619 | 152 | |
NM_018995.2 | NP_061868.1 | 9 - 10 | 1542 | 152 | |
XM_005261923.3 | XP_005261980.1 | 9 - 10 | 1820 | 152 | |
XM_011530696.1 | XP_011528998.1 | 9 - 10 | 1542 | 152 | |
XM_011530697.1 | XP_011528999.1 | 9 - 10 | 1820 | 152 | |
XM_011530698.1 | XP_011529000.1 | 8 - 9 | 1401 | 152 | |
XM_011530699.1 | XP_011529001.1 | 9 - 10 | 1504 | 152 | |
XM_011530700.2 | XP_011529002.1 | 9 - 10 | 1887 | 152 | |
XM_011530701.1 | XP_011529003.1 | 9 - 10 | 1542 | 152 | |
XM_011530702.1 | XP_011529004.1 | 8 - 9 | 1315 | 152 | |
XM_011530704.1 | XP_011529006.1 | 9 - 10 | 1542 | 152 | |
XM_017028833.1 | XP_016884322.1 | 9 - 10 | 1542 | 152 | |
XM_017028834.1 | XP_016884323.1 | 9 - 10 | 1542 | 152 | |
XM_017028837.1 | XP_016884326.1 | 9 - 10 | 1542 | 152 | |
GenBank mRNA | AF285604.1 | - | 9 - 10 | 1486 | 152 |
AK092675.1 | - | 3 - 4 | 378 | 152 | |
AK302401.1 | - | 9 - 10 | 1619 | 152 | |
AK303017.1 | - | 4 - 5 | 756 | 152 | |
BC140944.1 | - | 10 - 11 | 1543 | 152 | |
BC150137.1 | - | 9 - 10 | 1484 | 152 | |
BC152539.1 | - | 9 - 10 | 1484 | 152 | |
CR456466.1 | - | 9 - 10 | 1465 | 152 |