Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBLN1
Entrez Gene ID
2192
Gene Name
fibulin 1
Gene Aliases
FBLN, FIBL1
Chromosome Location
Chr.22: 45502839 - 45601134 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001996.3 NP_001987.2 2 - 3 334 107
NM_006485.3 NP_006476.2 2 - 3 334 107
NM_006486.2 NP_006477.2 2 - 3 334 107
NM_006487.2 NP_006478.2 2 - 3 334 107
GenBank mRNA AF126110.1 - 2 - 3 197 107
AF217999.1 - 2 - 3 276 107
AK075566.1 - 2 - 3 293 107
AK300806.1 - 2 - 3 270 107
AK316291.1 - 2 - 3 270 107
BC022497.1 - 2 - 3 290 107
BX424750.2 - 2 - 3 332 107
U01244.1 - 2 - 3 197 107
X53741.1 - 2 - 3 197 107
X53742.1 - 2 - 3 197 107
X53743.1 - 2 - 3 197 107

View Full Product Details