The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014646.2 | NP_055461.1 | 5 - 6 | 936 | 82 |
XM_005258177.4 | XP_005258234.1 | 5 - 6 | 1006 | 82 | |
XM_005258178.3 | XP_005258235.1 | 5 - 6 | 935 | 82 | |
XM_005258179.4 | XP_005258236.1 | 5 - 6 | 869 | 82 | |
XM_017026098.1 | XP_016881587.1 | 6 - 7 | 1025 | 82 | |
XM_017026099.1 | XP_016881588.1 | 5 - 6 | 796 | 82 | |
GenBank mRNA | BC152448.1 | - | 5 - 6 | 936 | 82 |
D87436.1 | - | 5 - 6 | 936 | 82 |