Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNRNPUL1
Entrez Gene ID
11100
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases
E1B-AP5, E1BAP5, HNRPUL1
Chromosome Location
Chr.19: 41262476 - 41307783 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301016.2 NP_001287945.1 6 - 7 885 86
NM_001321208.1 NP_001308137.1 6 - 7 925 86
NM_001321211.1 NP_001308140.1 6 - 7 709 86
NM_007040.5 NP_008971.2 6 - 7 1216 86
NM_144732.4 NP_653333.1 6 - 7 698 86
XM_005258459.3 XP_005258516.1 6 - 7 1019 86
XM_005258461.4 XP_005258518.1 6 - 7 849 86
XM_005258463.3 XP_005258520.1 6 - 7 840 86
XM_005258464.3 XP_005258521.1 6 - 7 655 86
XM_005258465.4 XP_005258522.1 6 - 7 866 86
XM_011526392.2 XP_011524694.1 6 - 7 1019 86
XM_011526393.2 XP_011524695.1 6 - 7 941 86
XM_011526394.2 XP_011524696.1 6 - 7 696 86
XM_011526395.2 XP_011524697.1 6 - 7 905 86
GenBank mRNA AJ007509.1 - 6 - 7 1059 86
AK022863.1 - 6 - 7 1018 86
AK023006.1 - 6 - 7 655 86
AK225850.1 - 6 - 7 678 86
AK290729.1 - 6 - 7 680 86
AK291315.1 - 7 - 8 965 86
AK291762.1 - 6 - 7 1018 86
AK297127.1 - 6 - 7 813 86
AK308078.1 - 6 - 7 946 86
AK308156.1 - 7 - 8 698 86
BC002564.2 - 6 - 7 956 86
BC009988.2 - 6 - 7 862 86
BC014232.2 - 6 - 7 837 86
BC027713.2 - 6 - 7 939 86

View Full Product Details