Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCST2
Entrez Gene ID
127579
Gene Name
DC-STAMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 155018513 - 155033781 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144622.2 NP_653223.2 5 - 6 888 58
XM_011509188.2 XP_011507490.1 3 - 4 351 58
GenBank mRNA AK057496.1 - 5 - 6 888 58
AK097468.1 - 3 - 4 302 58
BC111794.1 - 5 - 6 867 58

View Full Product Details