The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287816.1 | NP_001274745.1 | 26 - 27 | 3369 | 75 |
XM_005245670.3 | XP_005245727.1 | 26 - 27 | 3372 | 75 | |
XM_005245672.3 | XP_005245729.1 | 26 - 27 | 3261 | 75 | |
XM_017002985.1 | XP_016858474.1 | 26 - 27 | 3397 | 75 | |
XM_017002986.1 | XP_016858475.1 | 26 - 27 | 3264 | 75 | |
XM_017002987.1 | XP_016858476.1 | 26 - 27 | 3295 | 75 | |
XM_017002988.1 | XP_016858477.1 | 26 - 27 | 3292 | 75 | |
GenBank mRNA | AJ243670.2 | - | 10 - 11 | 1163 | 75 |
AK302662.1 | - | 26 - 27 | 3303 | 75 |