The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012114.2 | NP_036246.1 | 1 - 2 | 169 | 90 |
XM_011527861.1 | XP_011526163.1 | 1 - 2 | 169 | 90 | |
GenBank mRNA | AK313992.1 | - | 1 - 2 | 169 | 90 |
EU502748.1 | - | 1 - 2 | 265 | 90 |