The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012114.2 | NP_036246.1 | 5 - 6 | 734 | 111 |
GenBank mRNA | AF097874.1 | - | 4 - 5 | 570 | 111 |
AK313992.1 | - | 5 - 6 | 734 | 111 | |
BC069541.1 | - | 4 - 5 | 570 | 111 | |
BC103868.1 | - | 4 - 5 | 570 | 111 | |
BC103869.1 | - | 4 - 5 | 570 | 111 | |
EU262664.1 | - | 4 - 5 | 570 | 111 | |
EU502748.1 | - | 5 - 6 | 830 | 111 |