The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297549.1 | NP_001284478.1 | 20 - 21 | 3164 | 69 |
NM_002911.3 | NP_002902.2 | 20 - 21 | 3131 | 69 | |
XM_017027105.1 | XP_016882594.1 | 20 - 21 | 3197 | 69 | |
XM_017027106.1 | XP_016882595.1 | 20 - 21 | 3164 | 69 | |
GenBank mRNA | AF074016.1 | - | 20 - 21 | 3083 | 69 |
AK125797.1 | - | 2 - 3 | 761 | 69 | |
AK128727.1 | - | 5 - 6 | 1613 | 69 | |
BC039817.1 | - | 20 - 21 | 3128 | 69 | |
D86988.1 | - | 20 - 21 | 3098 | 69 | |
U59323.1 | - | 20 - 21 | 3031 | 69 | |
U65533.1 | - | 20 - 21 | 3087 | 69 |