The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297549.1 | NP_001284478.1 | 4 - 5 | 903 | 85 |
NM_002911.3 | NP_002902.2 | 4 - 5 | 903 | 85 | |
XM_017027105.1 | XP_016882594.1 | 4 - 5 | 912 | 85 | |
XM_017027106.1 | XP_016882595.1 | 4 - 5 | 912 | 85 | |
GenBank mRNA | AF074016.1 | - | 4 - 5 | 855 | 85 |
BC039817.1 | - | 4 - 5 | 900 | 85 | |
D86988.1 | - | 4 - 5 | 837 | 85 | |
U59323.1 | - | 4 - 5 | 803 | 85 | |
U65533.1 | - | 4 - 5 | 859 | 85 |