The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297549.1 | NP_001284478.1 | 7 - 8 | 1367 | 92 |
XM_017027105.1 | XP_016882594.1 | 7 - 8 | 1376 | 92 | |
GenBank mRNA | D86988.1 | - | 7 - 8 | 1301 | 92 |