The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005071.2 | NP_005062.1 | 8 - 9 | 1510 | 74 |
NR_073589.1 | 8 - 9 | 1463 | 74 | ||
XM_006722842.1 | XP_006722905.1 | 9 - 10 | 1693 | 74 | |
XM_006722844.1 | XP_006722907.1 | 9 - 10 | 1586 | 74 | |
XM_017027152.1 | XP_016882641.1 | 9 - 10 | 1781 | 74 | |
GenBank mRNA | AK302380.1 | - | 8 - 9 | 1440 | 74 |
AK312996.1 | - | 8 - 9 | 1542 | 74 | |
U18244.1 | - | 8 - 9 | 1510 | 74 |