The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000138.4 | NP_000129.3 | 24 - 25 | 3250 | 100 |
GenBank mRNA | BC146854.1 | - | 24 - 25 | 3116 | 100 |
L13923.1 | - | 23 - 24 | 2988 | 100 | |
X62008.1 | - | 4 - 5 | 467 | 100 | |
X63556.1 | - | 24 - 25 | 3250 | 100 |