Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF23
Entrez Gene ID
9493
Gene Name
kinesin family member 23
Gene Aliases
CHO1, KNSL5, MKLP-1, MKLP1
Chromosome Location
Chr.15: 69414246 - 69448427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281301.1 NP_001268230.1 10 - 11 1240 111
NM_004856.6 NP_004847.2 11 - 12 1377 111
NM_138555.3 NP_612565.1 11 - 12 1377 111
XM_005254796.2 XP_005254853.1 12 - 13 1368 111
XM_005254797.2 XP_005254854.1 12 - 13 1368 111
XM_005254798.2 XP_005254855.1 12 - 13 1368 111
XM_005254799.2 XP_005254856.1 12 - 13 1368 111
XM_011522238.1 XP_011520540.1 11 - 12 1203 111
XM_011522239.2 XP_011520541.1 11 - 12 1228 111
XM_011522240.1 XP_011520542.1 3 - 4 222 111
XM_017022733.1 XP_016878222.1 12 - 13 1368 111
GenBank mRNA AK303874.1 - 7 - 8 771 111
AK308322.1 - 10 - 11 1162 111
AK308361.1 - 5 - 6 586 111
AK310402.1 - 13 - 14 1446 111
BC017705.2 - 10 - 11 1145 111
BC051826.1 - 11 - 12 1319 111
BX477359.1 - 2 - 3 161 111
X67155.2 - 10 - 11 1217 111

View Full Product Details