The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281301.1 | NP_001268230.1 | 15 - 16 | 1972 | 74 |
NM_004856.6 | NP_004847.2 | 16 - 17 | 2109 | 74 | |
NM_138555.3 | NP_612565.1 | 16 - 17 | 2109 | 74 | |
XM_005254796.2 | XP_005254853.1 | 17 - 18 | 2100 | 74 | |
XM_005254797.2 | XP_005254854.1 | 17 - 18 | 2100 | 74 | |
XM_005254798.2 | XP_005254855.1 | 17 - 18 | 2100 | 74 | |
XM_005254799.2 | XP_005254856.1 | 17 - 18 | 2100 | 74 | |
XM_011522238.1 | XP_011520540.1 | 16 - 17 | 1935 | 74 | |
XM_011522239.2 | XP_011520541.1 | 16 - 17 | 1960 | 74 | |
XM_011522240.1 | XP_011520542.1 | 8 - 9 | 954 | 74 | |
GenBank mRNA | AK303874.1 | - | 12 - 13 | 1503 | 74 |
AK308322.1 | - | 15 - 16 | 1894 | 74 | |
AK308361.1 | - | 10 - 11 | 1318 | 74 | |
BC017705.2 | - | 15 - 16 | 1877 | 74 | |
BC051826.1 | - | 16 - 17 | 2051 | 74 | |
BX327654.2 | - | 1 - 2 | 104 | 74 | |
X67155.2 | - | 15 - 16 | 1949 | 74 |