The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281301.1 | NP_001268230.1 | 18 - 19 | 2544 | 96 |
NM_004856.6 | NP_004847.2 | 19 - 20 | 2621 | 96 | |
NM_138555.3 | NP_612565.1 | 20 - 21 | 2933 | 96 | |
XM_005254796.2 | XP_005254853.1 | 21 - 22 | 2924 | 96 | |
XM_005254798.2 | XP_005254855.1 | 20 - 21 | 2672 | 96 | |
XM_005254799.2 | XP_005254856.1 | 20 - 21 | 2612 | 96 | |
XM_011522238.1 | XP_011520540.1 | 20 - 21 | 2759 | 96 | |
XM_011522239.2 | XP_011520541.1 | 20 - 21 | 2784 | 96 | |
XM_011522240.1 | XP_011520542.1 | 12 - 13 | 1778 | 96 | |
XM_017022733.1 | XP_016878222.1 | 18 - 19 | 2258 | 96 | |
GenBank mRNA | AK303874.1 | - | 15 - 16 | 2015 | 96 |
AK308322.1 | - | 18 - 19 | 2466 | 96 | |
BC017705.2 | - | 19 - 20 | 2701 | 96 | |
BC051826.1 | - | 19 - 20 | 2563 | 96 | |
BX327654.2 | - | 4 - 5 | 616 | 96 | |
X67155.2 | - | 18 - 19 | 2461 | 96 |