The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018089.2 | NP_001018099.1 | 6 - 7 | 756 | 133 |
NM_001276385.1 | NP_001263314.1 | 6 - 7 | 916 | 133 | |
NM_024611.5 | NP_078887.2 | 6 - 7 | 916 | 133 | |
XM_005254661.1 | XP_005254718.1 | 7 - 8 | 915 | 133 | |
XM_005254662.1 | XP_005254719.1 | 7 - 8 | 778 | 133 | |
XM_011522009.1 | XP_011520311.1 | 6 - 7 | 790 | 133 | |
XM_011522010.1 | XP_011520312.1 | 6 - 7 | 659 | 133 | |
XM_011522011.2 | XP_011520313.1 | 5 - 6 | 573 | 133 | |
XM_017022569.1 | XP_016878058.1 | 7 - 8 | 952 | 133 | |
GenBank mRNA | AF502591.1 | - | 6 - 7 | 859 | 133 |
AK055752.1 | - | 6 - 7 | 859 | 133 | |
AK095389.1 | - | 6 - 7 | 740 | 133 | |
AL832046.1 | - | 6 - 7 | 883 | 133 | |
AY358546.1 | - | 6 - 7 | 877 | 133 | |
BC020918.1 | - | 6 - 7 | 1120 | 133 | |
BC140894.1 | - | 6 - 7 | 835 | 133 | |
BC151844.1 | - | 6 - 7 | 830 | 133 | |
BX647873.1 | - | 6 - 7 | 742 | 133 | |
CR749852.1 | - | 6 - 7 | 709 | 133 |