The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018009.4 | NP_060479.3 | 4 - 5 | 1144 | 63 |
XM_005253700.3 | XP_005253757.1 | 4 - 5 | 1144 | 63 | |
XM_017019545.1 | XP_016875034.1 | 4 - 5 | 1292 | 63 | |
XM_017019546.1 | XP_016875035.1 | 4 - 5 | 972 | 63 | |
GenBank mRNA | AK001005.1 | - | 4 - 5 | 1048 | 63 |
AK002056.2 | - | 4 - 5 | 1032 | 63 | |
AK098035.1 | - | 5 - 6 | 1106 | 63 | |
AK222956.1 | - | 4 - 5 | 1032 | 63 | |
AK223163.1 | - | 4 - 5 | 1062 | 63 | |
BC015017.2 | - | 4 - 5 | 1071 | 63 |