Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7orf26
Entrez Gene ID
79034
Gene Name
chromosome 7 open reading frame 26
Gene Aliases
-
Chromosome Location
Chr.7: 6589368 - 6608726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303039.1 NP_001289968.1 3 - 4 792 90
NM_024067.3 NP_076972.2 3 - 4 849 90
XM_005249845.4 XP_005249902.1 3 - 4 867 90
XM_011515518.2 XP_011513820.1 3 - 4 867 90
XM_017012614.1 XP_016868103.1 3 - 4 1395 90
GenBank mRNA AK056149.1 - 3 - 4 586 90
BC001076.2 - 3 - 4 668 90
BC005121.1 - 3 - 4 668 90

View Full Product Details