Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UHRF2
Entrez Gene ID
115426
Gene Name
ubiquitin like with PHD and ring finger domains 2
Gene Aliases
NIRF, RNF107, TDRD23, URF2
Chromosome Location
Chr.9: 6413148 - 6508108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152896.2 NP_690856.1 13 - 14 2345 83
NR_046386.1 14 - 15 2444 83
XM_011517704.2 XP_011516006.1 13 - 14 1974 83
XM_011517705.2 XP_011516007.1 11 - 12 1473 83
XM_017014253.1 XP_016869742.1 12 - 13 2129 83
XM_017014254.1 XP_016869743.1 14 - 15 2077 83
GenBank mRNA AB071698.1 - 13 - 14 2043 83
AF274049.1 - 13 - 14 2173 83
AL137728.1 - 1 - 2 1580 83
AL833522.1 - 7 - 8 2180 83
BC028397.2 - 14 - 15 2444 83

View Full Product Details