The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130475.2 | NP_001123947.1 | 1 - 2 | 277 | 115 |
GenBank mRNA | AL833137.1 | - | 1 - 2 | 232 | 115 |