The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190906.1 | NP_001177835.1 | 3 - 4 | 470 | 132 |
NM_001190907.1 | NP_001177836.1 | 3 - 4 | 470 | 132 | |
NM_005989.3 | NP_005980.1 | 3 - 4 | 470 | 132 | |
GenBank mRNA | AK058142.1 | - | 3 - 4 | 293 | 132 |
AK289425.1 | - | 3 - 4 | 410 | 132 | |
AK298421.1 | - | 3 - 4 | 471 | 132 | |
AK298428.1 | - | 3 - 4 | 436 | 132 | |
BC130625.1 | - | 3 - 4 | 425 | 132 | |
BC130627.1 | - | 3 - 4 | 425 | 132 | |
FM160741.1 | - | 6 - 7 | 966 | 132 | |
FM160742.1 | - | 5 - 6 | 804 | 132 | |
Z28339.1 | - | 3 - 4 | 452 | 132 |