Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNASE8
Entrez Gene ID
122665
Gene Name
ribonuclease A family member 8
Gene Aliases
RAE2
Chromosome Location
Chr.14: 21057822 - 21058455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138331.2 NP_612204.1 1 - 1 486 72
GenBank mRNA BC137537.1 - 1 - 1 486 72
BC137538.1 - 1 - 1 486 72

View Full Product Details