Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOGL
Entrez Gene ID
283310
Gene Name
otogelin like
Gene Aliases
C12orf64, DFNB84B
Chromosome Location
Chr.12: 80130377 - 80379090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173591.3 NP_775862.3 36 - 37 4327 96
XM_005268802.3 XP_005268859.1 40 - 41 5128 96
XM_011538191.1 XP_011536493.1 37 - 38 4404 96
XM_011538192.2 XP_011536494.1 34 - 35 4345 96
XM_011538193.1 XP_011536495.1 34 - 35 4264 96

View Full Product Details