The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173591.3 | NP_775862.3 | 33 - 34 | 4057 | 120 |
XM_005268802.3 | XP_005268859.1 | 37 - 38 | 4858 | 120 | |
XM_011538191.1 | XP_011536493.1 | 34 - 35 | 4134 | 120 | |
XM_011538192.2 | XP_011536494.1 | 31 - 32 | 4075 | 120 | |
XM_011538193.1 | XP_011536495.1 | 31 - 32 | 3994 | 120 |