The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014502.4 | NP_055317.1 | 15 - 16 | 1621 | 88 |
GenBank mRNA | BC008719.2 | - | 15 - 16 | 1616 | 88 |
BC018665.2 | - | 15 - 16 | 1616 | 88 | |
BC018698.2 | - | 15 - 16 | 1616 | 88 |