The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006392.3 | NP_006383.2 | 10 - 11 | 1410 | 81 |
NR_027700.2 | 10 - 11 | 1590 | 81 | ||
GenBank mRNA | AK292721.1 | - | 10 - 11 | 1310 | 81 |
BC004937.1 | - | 3 - 4 | 370 | 81 | |
BC018421.1 | - | 9 - 10 | 1270 | 81 | |
BC035369.1 | - | 10 - 11 | 1300 | 81 | |
BC104791.1 | - | 10 - 11 | 1410 | 81 | |
BC104793.1 | - | 10 - 11 | 1410 | 81 | |
BX641071.1 | - | 1 - 2 | 1350 | 81 | |
BX647439.1 | - | 10 - 11 | 1310 | 81 | |
BX647513.1 | - | 9 - 10 | 1334 | 81 | |
Y12065.1 | - | 9 - 10 | 87 |