The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207047.2 | NP_001193976.1 | 10 - 11 | 1499 | 95 |
NM_001323332.1 | NP_001310261.1 | 10 - 11 | 1882 | 95 | |
NM_139321.2 | NP_647537.1 | 10 - 11 | 1882 | 95 | |
NM_139322.3 | NP_647538.1 | 10 - 11 | 1882 | 95 | |
GenBank mRNA | AF106861.1 | - | 10 - 11 | 1792 | 95 |
AK293010.1 | - | 10 - 11 | 1882 | 95 | |
BC101705.1 | - | 10 - 11 | 1832 | 95 |