The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323332.1 | NP_001310261.1 | 1 - 2 | 500 | 110 |
NM_139321.2 | NP_647537.1 | 1 - 2 | 500 | 110 | |
NM_139322.3 | NP_647538.1 | 1 - 2 | 500 | 110 | |
GenBank mRNA | AF106861.1 | - | 1 - 2 | 410 | 110 |
AK293010.1 | - | 1 - 2 | 500 | 110 | |
BC101705.1 | - | 1 - 2 | 450 | 110 | |
DB240822.1 | - | 1 - 2 | 501 | 110 |