The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170765.1 | NP_001164236.1 | 1 - 2 | 139 | 132 |
NM_001170766.1 | NP_001164237.1 | 1 - 2 | 139 | 132 | |
XM_017016785.1 | XP_016872274.1 | 1 - 2 | 149 | 197 | |
XM_017016787.1 | XP_016872276.1 | 1 - 2 | 140 | 132 | |
GenBank mRNA | AL832106.1 | - | 1 - 2 | 119 | 132 |
BC053359.1 | - | 1 - 2 | 80 | 132 |