Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCOR
Entrez Gene ID
84458
Gene Name
ligand dependent nuclear receptor corepressor
Gene Aliases
MLR2
Chromosome Location
Chr.10: 96832250 - 96964441 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170765.1 NP_001164236.1 3 - 4 279 142
NM_001170766.1 NP_001164237.1 3 - 4 279 142
NM_032440.3 NP_115816.2 3 - 4 299 142
XM_006718033.3 XP_006718096.1 3 - 4 587 142
XM_011540277.2 XP_011538579.1 3 - 4 587 142
XM_017016784.1 XP_016872273.1 2 - 3 210 142
XM_017016785.1 XP_016872274.1 4 - 5 354 142
XM_017016786.1 XP_016872275.1 3 - 4 275 142
XM_017016788.1 XP_016872277.1 2 - 3 201 142
GenBank mRNA AB058698.1 - 3 - 4 212 142
AL832106.1 - 3 - 4 259 142
AL834245.2 - 3 - 4 282 142
BC053359.1 - 3 - 4 220 142
DA507116.1 - 3 - 4 299 142

View Full Product Details