The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024712.3 | NP_078988.2 | 10 - 11 | 1123 | 125 |
GenBank mRNA | AK023886.1 | - | 9 - 10 | 887 | 125 |
AK056923.1 | - | 9 - 10 | 1086 | 125 | |
AK300976.1 | - | 10 - 11 | 1137 | 125 | |
BC034410.1 | - | 7 - 8 | 735 | 125 |